• USA Home
  • 65680 - (S)-1-{(SP)-2-[2-(Diphenylphosphino)phenyl]ferrocenyl}ethyldi(3,5-xylyl)phosphine

EMAIL THIS PAGE TO A FRIEND
65680 Sigma-Aldrich

(S)-1-{(SP)-2-[2-(Diphenylphosphino)phenyl]ferrocenyl}ethyldi(3,5-xylyl)phosphine

≥97%

Synonym: (1R)-1-[(1S)-1-[Bis(3,5-dimethylphenyl)phosphino]ethyl]-2-[2-(diphenylphosphino)phenyl]ferrocene (acc to CAS), Walphos SL-W006-2

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Privileged Ligands and Complexes, Solvias Ligands and Complexes,
assay   ≥97%
optical purity   ee: ≥99%
storage temp.   room temp
SMILES string   [Fe].[CH]1[CH][CH][CH][CH]1.C[C@@H]([C]2[CH][CH][CH][C]2c3ccccc3P(c4ccccc4)c5ccccc5)P(c6cc(C)cc(C)c6)c7cc(C)cc(C)c7
InChI   1S/C41H39P2.C5H5.Fe/c1-29-23-30(2)26-36(25-29)42(37-27-31(3)24-32(4)28-37)33(5)38-20-14-21-39(38)40-19-12-13-22-41(40)43(34-15-8-6-9-16-34)35-17-10-7-11-18-35;1-2-4-5-3-1;/h6-28,33H,1-5H3;1-5H;/t33-;;/m0../s1
InChI key   SPJXDOOLGLNVOP-NYPSMHOZSA-N

Description

General description

sold in collaboration with Solvias AG

Packaging

1, 5 g in glass bottle

100, 500 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?