• USA Home
  • 65683 - (R)-1-{(RP)-2-[2-[Di(3,5-xylyl)phosphino]phenyl]ferrocenyl}ethyldi(3,5-xylyl)phosphine

EMAIL THIS PAGE TO A FRIEND
65683 Sigma-Aldrich

(R)-1-{(RP)-2-[2-[Di(3,5-xylyl)phosphino]phenyl]ferrocenyl}ethyldi(3,5-xylyl)phosphine

≥97%

Synonym: (1S)-1-[(1R)-1-[Bis(3,5-dimethylphenyl)phosphino]ethyl]-2-[2-[bis(3,5-dimethylphenyl)phosphino]phenyl]ferrocene (acc to CAS), Walphos SL-W009-1

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Privileged Ligands and Complexes, Solvias Ligands and Complexes,
assay   ≥97%
optical purity   ee: ≥99%
storage temp.   room temp
SMILES string   [Fe].[CH]1[CH][CH][CH][CH]1.C[C@@H]([C]2[CH][CH][CH][C]2c3ccccc3P(c4cc(C)cc(C)c4)c5cc(C)cc(C)c5)P(c6cc(C)cc(C)c6)c7cc(C)cc(C)c7
InChI   1S/C45H47P2.C5H5.Fe/c1-29-17-30(2)22-38(21-29)46(39-23-31(3)18-32(4)24-39)37(9)42-14-12-15-43(42)44-13-10-11-16-45(44)47(40-25-33(5)19-34(6)26-40)41-27-35(7)20-36(8)28-41;1-2-4-5-3-1;/h10-28,37H,1-9H3;1-5H;/t37-;;/m0../s1
InChI key   IDRVINOGJFWZOP-SHRURHRBSA-N

Description

General description

sold in collaboration with Solvias AG

Packaging

1, 5 g in glass bottle

100, 500 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Solvias Chiral Phosphine Ligands - Toolkit for Asymmetric Catalysis

Sigma-Aldrich, in collaboration with Solvias, is proud to present the Chiral Ligands Kit – the ultimate toolkit for asymmetric catalysis!
Keywords: Asymmetric catalysis, Asymmetric synthesis, Catalysis, Individual protein Immunoprecipitation, Ligands

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?