• USA Home
  • 657050 - Potassium 4-acetylphenyltrifluoroborate

EMAIL THIS PAGE TO A FRIEND
657050 Sigma-Aldrich

Potassium 4-acetylphenyltrifluoroborate

97%

Purchase

Properties

Related Categories Aryl Trifluoroborate Salts, Boronic Acids and Derivatives, Chemical Synthesis, Organometallic Reagents, Trifluoroborate Salts More...
assay   97%
mp   290 °C (dec.)
SMILES string   [K+].CC(=O)c1ccc(cc1)[B-](F)(F)F
InChI   1S/C8H7BF3O.K/c1-6(13)7-2-4-8(5-3-7)9(10,11)12;/h2-5H,1H3;/q-1;+1
InChI key   JRUXIRSHKHCTTK-UHFFFAOYSA-N

Description

Packaging

1, 5 g in poly bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Potassium Trifluoroborate Salts

Boronic acids, boronate esters, and organoboranes have been utilized for many years as the primary boron source in Suzuki–Miyaura-type reactions. These organoboron reagents have inherent limitations ...
ChemFiles Volume 5 Article 10
Keywords: Aldrichimica Acta, Coupling reactions, Cross couplings, Degradations, Epoxidations, Type

Professor and Product Portal - Gary Molander Group

From our library of Articles, Sigma-Aldrich presents Professor and Product Portal - Gary Molander Group
Keywords: Catalysis, Coupling reactions, Cross couplings, Transmetalation

Stabilized 2-Iodoxybenzoic Acid (SIBX)

Since 1994,1 2-iodoxybenzoic acid (IBX) has been well recognized as a very powerful and selective oxidizing agent. Similar to the Dess–Martin periodinane, IBX is an environmentally benign alternative...
ChemFiles 2006, 6.2, 14.
Keywords: Dearomatizations, Oxidations

Related Content

Organotrifluoroborates in Chemical Synthesis

Potent Boronic Acid Surrogates In the vast array of catalytic carbon-carbon bond forming reactions, the Suzuki-Miyaura protocol has proven to be a particularly attractive method.  The organoborane nu...
Keywords: Addition reactions, Catalysis, Coupling reactions, Cross couplings, Halogenations, Suzuki-Miyaura coupling

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?