• USA Home
  • 657115 - Potassium 4-(methylthio)phenyltrifluoroborate

EMAIL THIS PAGE TO A FRIEND
657115 Sigma-Aldrich

Potassium 4-(methylthio)phenyltrifluoroborate

95%

Purchase

Properties

Related Categories Aryl Trifluoroborate Salts, Boronic Acids and Derivatives, Chemical Synthesis, Organometallic Reagents, Trifluoroborate Salts More...
assay   95%
mp   >300 °C
SMILES string   [K+].CSc1ccc(cc1)[B-](F)(F)F
InChI   1S/C7H7BF3S.K/c1-12-7-4-2-6(3-5-7)8(9,10)11;/h2-5H,1H3;/q-1;+1
InChI key   RXGCQQOLNQOSLX-UHFFFAOYSA-N

Description

Packaging

1, 5 g in poly bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Organotrifluoroborates in Chemical Synthesis

Potent Boronic Acid Surrogates In the vast array of catalytic carbon-carbon bond forming reactions, the Suzuki-Miyaura protocol has proven to be a particularly attractive method.  The organoborane nu...
Keywords: Addition reactions, Catalysis, Coupling reactions, Cross couplings, Halogenations, Suzuki-Miyaura coupling

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?