• USA Home
  • 657387 - 4-(2′-Methoxybenzyloxy)phenylboronic acid

EMAIL THIS PAGE TO A FRIEND
657387 Sigma-Aldrich

4-(2′-Methoxybenzyloxy)phenylboronic acid

Purchase

Properties

Related Categories Aryl Boronic Acids, Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Monosubstituted Aryl Boronic Acids,
InChI Key   YPZFEBLXRQPHIO-UHFFFAOYSA-N
mp   172-180 °C

Description

Packaging

10 g in glass bottle

Other Notes

Contains varying amounts of anhydride

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

28982 Curcumin solution, ~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v)), for TLC derivatization

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?