• USA Home
  • 657417 - 2-(4-Methoxybenzyloxy)phenylboronic acid

EMAIL THIS PAGE TO A FRIEND
657417 Sigma-Aldrich

2-(4-Methoxybenzyloxy)phenylboronic acid

≥95%

  • Empirical Formula (Hill Notation) C14H15BO4

  • Molecular Weight 258.08

  •  MDL number MFCD07784391

  •  PubChem Substance ID 24884287

Purchase

Properties

Related Categories Aryl Boronic Acids, Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Monosubstituted Aryl Boronic Acids,
InChI Key   YVWFATUNDBQXOZ-UHFFFAOYSA-N
assay   ≥95%
mp   112-116 °C

Description

Packaging

2, 10 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?