• USA Home
  • 657522 - 4-[(1-Naphthyloxy)methyl]phenylboronic acid

EMAIL THIS PAGE TO A FRIEND
657522 Sigma-Aldrich

4-[(1-Naphthyloxy)methyl]phenylboronic acid

≥95%

Purchase

Properties

Related Categories Aryl Boronic Acids, Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Monosubstituted Aryl Boronic Acids,
InChI Key   ZYQMTIWKJLAEEF-UHFFFAOYSA-N
assay   ≥95%
mp   206-220 °C

Description

Packaging

2 g in glass bottle

Other Notes

Contains varying amounts of anhydride

Safety & Documentation

Safety Information

Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?