• USA Home
  • 657549 - 1-Indanone-6-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
657549 Sigma-Aldrich

1-Indanone-6-carboxylic acid

97%

Synonym: 2,3-Dihydro-3-oxo-1H-indene-5-carboxylic acid, 3-Oxoindane-5-carboxylic acid, 6-Carboxy-1-indanone

Purchase

Properties

Related Categories Building Blocks, C10, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
assay   97%
mp   256-260 °C
SMILES string   OC(=O)c1ccc2CCC(=O)c2c1
InChI   1S/C10H8O3/c11-9-4-3-6-1-2-7(10(12)13)5-8(6)9/h1-2,5H,3-4H2,(H,12,13)
InChI key   BIABIACQHKYEEB-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?