• USA Home
  • 659169 - [6,6]-Phenyl C61 butyric acid methyl ester

EMAIL THIS PAGE TO A FRIEND
659169 Aldrich

[6,6]-Phenyl C61 butyric acid methyl ester

Synonym: 1-[3-(Methoxycarbonyl)propyl]-1-phenyl-[6.6]C61, 3′H-Cyclopropa[1,9] [5,6]fullerene-C60-Ih-3′-butanoic acid 3′-phenyl methyl ester, PCBM, [60]PCBM

Purchase

Properties

description   functionalized fullerene

Description

Application

Soluble n-channel organic semiconductor. For use as an n-type layer in plastic electronics, especially bulk heterojunction OFETs and photovoltaic cells (PVs).

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Functionalized Fullerenes: Nanomaterials for Organic Electronics

Fullerenes, the older nanomaterial relatives of carbon nanotubes, continue to stimulate advances in applied and fundamental science. Fullerenes are excellent electron acceptors and can be chemically ...
Material Matters 2007, 2.1, 18.
Keywords: Electronics, Nanotubes, Organic electronics, Semiconductor, Solvents

Related Products

comparable product

Product #

Description

Add to Cart

684449 [6,6]-Phenyl C61 butyric acid methyl ester, >99.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?