• USA Home
  • 659428 - QuadraPure® Metal Scavengers Kit I

EMAIL THIS PAGE TO A FRIEND
659428 Aldrich

QuadraPure® Metal Scavengers Kit I

Purchase

Description

Application

Catalysis Screening Kits

Legal Information

QuadraPure is a registered trademark of Johnson Matthey Finland Oy

Kit component also available separately

Description

Product #

Add to Cart

QuadraPure® TU, macroporous, 400-600 μm particle size 5 g SDS 655422
QuadraPure® IDA, macroporous, 350-750 μm particle size 5 g SDS 657026
QuadraPure® AMPA, macroporous, 350-750 μm particle size 5 g SDS 657611
QuadraPure® AEA, 100-400 μm particle size, extent of labeling: 1.3 mmol/g loading, 1 % cross-linked with divinylbenzene 5 g SDS 657646
QuadraPure® IMDAZ, 100-400 μm particle size, extent of labeling: 1.5 mmol/g loading, 1 % cross-linked with divinylbenzene 5 g SDS 657654
QuadraPure® MPA, 100-400 μm particle size, extent of labeling: 1.5 mmol/g loading, 1 % cross-linked with divinylbenzene 5 g SDS 657662
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Catalysis Screening Kits

Accelerate methodology discovery and process optimization with Sigma‑Aldrich’s new chemistry kits for catalysis screening. Kits contain sets of related catalysts, ligands, and scavengers with varying...
Aldrich ChemFiles 2006, 6.2, 17.
Keywords: Catalysis, Chemfiles, Ligands

Related Content

Reaxa Technology

Easier, Faster, and Cleaner Reactions In partnership with Reaxa Ltd., Sigma-Aldrich is pleased to offer several technology platforms that allow for easier, faster, and cleaner processes. The technolo...
Keywords: Catalysis, Filtration, Metal scavengers, Methods, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?