• USA Home
  • 659886 - Yttrium trifluoroacetate

EMAIL THIS PAGE TO A FRIEND
659886 Sigma-Aldrich

Yttrium trifluoroacetate

≥99.99% trace metals basis, anhydrous

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals, Materials Science, Metal and Ceramic Science,
grade   anhydrous
InChI Key   NPATVYPXYMSPKD-UHFFFAOYSA-K
assay   ≥99.99% trace metals basis
form   powder

Description

Packaging

25 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?