• USA Home
  • 660442 - 3-Aminopropyl functionalized silica

EMAIL THIS PAGE TO A FRIEND
660442 Sigma-Aldrich

3-Aminopropyl functionalized silica

nanoparticles, <100 nm particle size (DLS), dispersion, 3 % (w/v) in ethanol

Purchase

Properties

Related Categories 14: Si, Materials Science, Nanomaterials, Nanopowders and Nanoparticle Dispersions, Surface Functionalized Nanoparticles More...
form   dispersion
concentration   3 % (w/v) in ethanol
particle size   <100 nm (DLS)
bp   75-78 °C
density   0.836 g/mL at 25 °C
functional group   amine
storage temp.   2-8°C

Description

Packaging

25 mL in glass bottle

Application

Nanoparticle building block for self-assembly.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1170 - class 3 - PG 2 - Ethanol, solution
WGK Germany 
3
Flash Point(F) 
39.2 °F
Flash Point(C) 
4 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?