• USA Home
  • 660612 - 2,3,5-Trifluoro-4-(trifluoromethyl)pyridine

EMAIL THIS PAGE TO A FRIEND
660612 Sigma-Aldrich

2,3,5-Trifluoro-4-(trifluoromethyl)pyridine

97%

  • CAS Number 675602-93-4

  • Empirical Formula (Hill Notation) C6HF6N

  • Molecular Weight 201.07

  •  MDL number MFCD03840202

  •  PubChem Substance ID 24884485

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, C6, Chemical Synthesis, Fluorinated Building Blocks, Heterocyclic Building Blocks,
assay   97%
refractive index   n20/D 1.383
bp   102-104 °C
density   1.576 g/mL at 25 °C
SMILES string   Fc1cnc(F)c(F)c1C(F)(F)F
InChI   1S/C6HF6N/c7-2-1-13-5(9)4(8)3(2)6(10,11)12/h1H
InChI key   MLFFFTHLIOLZFO-UHFFFAOYSA-N

Description

Packaging

100, 500 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1992 6.1(3) / PGIII
WGK Germany 
3
Flash Point(F) 
93.2 °F
Flash Point(C) 
34 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?