• USA Home
  • 66137 - Butyl 2-methylbutyrate

EMAIL THIS PAGE TO A FRIEND
66137 Sigma-Aldrich

Butyl 2-methylbutyrate

≥98.0%

  • CAS Number 15706-73-7

  • Linear Formula CH3CH2CH(CH3)COOCH2CH2CH2CH3

  • Molecular Weight 158.24

  •  Beilstein/REAXYS Number 1702551

  •  EC Number 239-798-2

  •  MDL number MFCD00042902

  •  PubChem Substance ID 57652048

Purchase

Properties

Related Categories Building Blocks, C8 to C9, Carbonyl Compounds, Chemical Synthesis, Esters,
assay   ≥98.0%
refractive index   n20/D 1.409 (lit.)
  n20/D 1.409
bp   173-176 °C/730 mmHg (lit.)
density   0.863 g/mL at 25 °C (lit.)
SMILES string   CCCCOC(=O)C(C)CC
InChI   1S/C9H18O2/c1-4-6-7-11-9(10)8(3)5-2/h8H,4-7H2,1-3H3
InChI key   OTKQNSSMCDLVQV-UHFFFAOYSA-N

Description

Packaging

25 mL in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3272 3 / PGIII
WGK Germany 
3
Flash Point(F) 
140 °F
Flash Point(C) 
60 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?