• USA Home
  • 662097 - 4-[(2-Isopropyl-5-methylphenoxy)methyl]phenylboronic acid

EMAIL THIS PAGE TO A FRIEND
662097 Sigma-Aldrich

4-[(2-Isopropyl-5-methylphenoxy)methyl]phenylboronic acid

Purchase

Properties

Related Categories Aryl Boronic Acids, Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Monosubstituted Aryl Boronic Acids,
InChI Key   FBZSTYQOBCWTEC-UHFFFAOYSA-N
mp   173-177 °C

Description

Packaging

2, 10 g in glass bottle

Other Notes

Contains varying amounts of anhydride

Safety & Documentation

Safety Information

Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?