• USA Home
  • 662216 - NanoThinks ACID16

EMAIL THIS PAGE TO A FRIEND
662216 Sigma-Aldrich

NanoThinks ACID16

Synonym: 16-Mercaptohexadecanoic acid solution

Purchase

Properties

concentration   5 mM in ethanol
density   0.787 g/mL at 25 °C
SMILES string   OC(=O)CCCCCCCCCCCCCCCS

Description

Legal Information

NanoThinks is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1170C 3 / PGII
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?