EMAIL THIS PAGE TO A FRIEND
66370 Sigma-Aldrich

Methylcyclohexanol

technical, mixture of isomers

Synonym: ‘Methylhexaline’

  • CAS Number 25639-42-3

  • Empirical Formula (Hill Notation) C7H14O

  • Molecular Weight 114.19

  •  Beilstein/REAXYS Number 1445998

  •  EC Number 247-152-6

  •  MDL number MFCD00001443

  •  PubChem Substance ID 24884650

Purchase

Properties

grade   technical
refractive index   n20/D 1.46
bp   171-173 °C (lit.)
density   0.92 g/mL at 20 °C (lit.)
SMILES string   CC1CCCCC1O
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2617 3 / PGIII
WGK Germany 
1
RTECS 
GW0175000
Flash Point(F) 
105.8 °F
Flash Point(C) 
41 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?