• USA Home
  • 666769 - 4-(4′-Methoxybenzyloxy)phenylboronic acid

EMAIL THIS PAGE TO A FRIEND
666769 Sigma-Aldrich

4-(4′-Methoxybenzyloxy)phenylboronic acid

≥95%

Purchase

Properties

Related Categories Aryl Boronic Acids, Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Monosubstituted Aryl Boronic Acids,
InChI Key   UCYGEGISTWJFFA-UHFFFAOYSA-N
assay   ≥95%
mp   174-178 °C

Description

General description

May contain varying amounts of anhydride

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?