• USA Home
  • 66765 - Methylenecyclopropane

EMAIL THIS PAGE TO A FRIEND
66765 Sigma-Aldrich

Methylenecyclopropane

≥95.0% (HPLC)

  • CAS Number 6142-73-0

  • Empirical Formula (Hill Notation) C4H6

  • Molecular Weight 54.09

  •  Beilstein/REAXYS Number 741861

  •  EC Number 228-142-0

  •  MDL number MFCD00042620

  •  PubChem Substance ID 57652062

Purchase

Properties

assay   ≥95.0% (HPLC)
bp   9-12 °C
density   0.80 g/mL at 20 °C (lit.)
storage temp.   −20°C
SMILES string   C=C1CC1
InChI   1S/C4H6/c1-4-2-3-4/h1-3H2
InChI key   XSGHLZBESSREDT-UHFFFAOYSA-N

Description

Other Notes

Sales restrictions may apply

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2037F 2.1
WGK Germany 
3
Flash Point(F) 
<6.8 °F
Flash Point(C) 
<-14 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?