• USA Home
  • 668249 - 2,4-Dimethoxybenzenesulfonyl chloride

EMAIL THIS PAGE TO A FRIEND
668249 Sigma-Aldrich

2,4-Dimethoxybenzenesulfonyl chloride

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonyl Halides, Sulfur Compounds More...
InChI Key   AYGZKRRIULCJKC-UHFFFAOYSA-N
assay   97%
mp   71-75 °C
storage temp.   2-8°C

Description

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?