• USA Home
  • 668362 - 2,5-Dichloro-3-methylthiophene

EMAIL THIS PAGE TO A FRIEND
668362 Sigma-Aldrich

2,5-Dichloro-3-methylthiophene

96%

Purchase

Properties

Related Categories Building Blocks, C4 to C6, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks,
InChI Key   BUWSFDLUTRHPBO-UHFFFAOYSA-N
assay   96%
refractive index   n20/D 1.555
density   1.331 g/mL at 25 °C
storage temp.   2-8°C

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
168.8 °F
Flash Point(C) 
76 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?