• USA Home
  • 669245 - 1-Phenyl-1-hexyn-3-ol

EMAIL THIS PAGE TO A FRIEND
669245 Sigma-Aldrich

1-Phenyl-1-hexyn-3-ol

97%

  • CAS Number 1817-51-2

  • Empirical Formula (Hill Notation) C12H14O

  • Molecular Weight 174.24

  •  MDL number MFCD00046687

  •  PubChem Substance ID 24885031

  •  NACRES NA.22

Purchase

Properties

Related Categories Alkynes, Building Blocks, Chemical Synthesis, Internal, Organic Building Blocks More...
assay   97%
refractive index   n20/D 1.549
density   0.949 g/mL at 25 °C
SMILES string   CCCC(O)C#Cc1ccccc1
InChI   1S/C12H14O/c1-2-6-12(13)10-9-11-7-4-3-5-8-11/h3-5,7-8,12-13H,2,6H2,1H3
InChI key   HGUIQUVFOYTZNC-UHFFFAOYSA-N

Description

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
230.0 °F - closed cup
Flash Point(C) 
> 110 °C - closed cup

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles

Related Content

Alkynes and Acetylenic Building Blocks

We list over 600 alkyne building blocks, including a wide array of propargyl alcohols, acetylenic boron reagents, and halogenated substrates for cross-coupling. Additionally, many of these are termin...
Keywords: Building blocks, Coupling reactions, Cross couplings

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?