• USA Home
  • 671118 - 2-Acetamido-2-deoxy-β-D-glucopyranosyl azide 3,4,6-triacetate

EMAIL THIS PAGE TO A FRIEND
671118 Sigma-Aldrich

2-Acetamido-2-deoxy-β-D-glucopyranosyl azide 3,4,6-triacetate

≥98.0% (HPLC)

Synonym: 2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl azide

Purchase

Properties

Related Categories Azides, Building Blocks, Carbohydrate Azides, Chemical Biology, Chemical Synthesis,
assay   ≥98.0% (HPLC)
optical activity   [α]/D -45.5±2.5°, c = 1 in chloroform
storage temp.   −20°C
SMILES string   CC(=O)N[C@H]1[C@@H](O[C@H](COC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O)N=[N+]=[N-]
InChI   1S/C14H20N4O8/c1-6(19)16-11-13(25-9(4)22)12(24-8(3)21)10(5-23-7(2)20)26-14(11)17-18-15/h10-14H,5H2,1-4H3,(H,16,19)/t10-,11-,12-,13-,14-/m1/s1
InChI key   RMCFMPMNMQZHSF-DHGKCCLASA-N

Description

Packaging

1 g in glass bottle

250 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?