• USA Home
  • 671126 - (R,R)-(+)-2-Amino-1,2-diphenylethanol

EMAIL THIS PAGE TO A FRIEND
671126 Sigma-Aldrich

(R,R)-(+)-2-Amino-1,2-diphenylethanol

≥99.5% (HPLC)

  • CAS Number 88082-66-0

  • Linear Formula C6H5CH(NH2)CH(C6H5)OH

  • Molecular Weight 213.28

  •  Beilstein/REAXYS Number 1212829

  •  MDL number MFCD03093885

  •  PubChem Substance ID 329760405

Purchase

Properties

Related Categories Amino Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
assay   ≥99.5% (HPLC)
optical activity   [α]/D +124±3°, c = 0.87 in ethanol
optical purity   enantiomeric excess: ≥99.0% (HPLC)
mp   116-119 °C
storage temp.   2-8°C
SMILES string   N[C@@H]([C@H](O)c1ccccc1)c2ccccc2
InChI   1S/C14H15NO/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13-14,16H,15H2/t13-,14-/m1/s1
InChI key   GEJJWYZZKKKSEV-ZIAGYGMSSA-N

Description

Packaging

500 mg in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?