• USA Home
  • 671886 - Boc-4-(Fmoc-amino)-L-phenylalanine

EMAIL THIS PAGE TO A FRIEND
671886 Sigma-Aldrich

Boc-4-(Fmoc-amino)-L-phenylalanine

≥98.0%

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Phenylalanine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   ZKSJJSOHPQQZHC-VWLOTQADSA-N
assay   ≥98.0%
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Unnatural Amino Acids - ChemFiles 2008, 8.7, 3.

Unnatural amino acids, the non-proteinogenic amino acids that either occur naturally or are chemically synthesized, are becoming more and more important as tools for modern drug discovery research. D...
Matthias Junkers
ChemFiles 2008, 8.7, 3.
Keywords: Absorption, Addition reactions, Alkylations, Asymmetric synthesis, Building blocks, Degradations, Drug discovery, Metabolism, Molecular probes, Peptidomimetics, Pharmaceutical

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?