• USA Home
  • 672246 - 2-(Dicyclohexylphosphino)-1-phenylindole

EMAIL THIS PAGE TO A FRIEND
672246 Sigma-Aldrich

2-(Dicyclohexylphosphino)-1-phenylindole

95% (NMR)

Synonym: cataCXium® PInCy

  • CAS Number 740815-36-5

  • Empirical Formula (Hill Notation) C26H32NP

  • Molecular Weight 389.51

  •  Beilstein/REAXYS Number 9951803

  •  MDL number MFCD06798303

  •  PubChem Substance ID 329760463

Purchase

Description

General description

sold in collaboration with Solvias AG

Application

Catalyst for:
• Stereoselective isomerization of terminal alkenes
• Reductive carbonylation of aryl and vinyl halides
• Heck reaction
• Cross-coupling reactions
• Buchwald-Hartwig amination

• Ligand for palladium-catalyzed amination of aryl and heteroaryl chlorides

Packaging

1, 5 g in glass bottle

Legal Information

WO 2004/101581

cataCXium is a registered trademark of Evonik Degussa GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

cataCXium® - Ligands and Complexes for Efficient Cross-Coupling Reactions

Cross-coupling reactions are an important class of catalytic transformations with applications in polymer science as well as in the fine chemicals and pharmaceutical industries. The use of aryl chlor...
Keywords: Aminations, Buchwald-Hartwig amination, Cancer, Catalysis, Coupling reactions, Cross couplings, Industries, Ligands, Pharmaceutical, Polymer science, Suzuki coupling, transformation

cataCXium®

Cross-coupling reactions are an important class of catalytic transformations with applications in polymer science as well as in the fine chemicals and pharmaceutical industries. The use of aryl chlor...
William Sommer
ChemFiles Volume 7 Article 10
Keywords: Aminations, Applications, Buchwald-Hartwig amination, Catalysis, Coupling reactions, Cross couplings, Industries, Ligands, Pharmaceutical, Polymer science, Suzuki coupling, transformation

Related Content

cataCXium® - Ligands and Complexes for Efficient Cross-Coupling Reactions

Cross-coupling reactions are an important class of catalytic transformations with applications in polymer science as well as in the fine chemicals and pharmaceutical industries. The use of aryl chlor...
Keywords: Aminations, Applications, Buchwald-Hartwig amination, Cancer, Catalysis, Coupling reactions, Cross couplings, Industries, Ligands, Pharmaceutical, Polymer science, Suzuki coupling, transformation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?