EMAIL THIS PAGE TO A FRIEND
672319 Sigma-Aldrich

Fmoc-Phe(2-Br)-OH

≥98.0% (HPLC)

Synonym: Fmoc-2-bromo-L-phenylalanine

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry | Peptide Synthesis, Phenylalanine More...
InChI Key   GRJPAUULVKPBHU-QFIPXVFZSA-N
assay   ≥98.0% (HPLC)

Description

Packaging

500 mg in poly tube

Safety & Documentation

Safety Information

Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Unnatural Amino Acids - ChemFiles 2008, 8.7, 3.

Unnatural amino acids, the non-proteinogenic amino acids that either occur naturally or are chemically synthesized, are becoming more and more important as tools for modern drug discovery research. D...
Matthias Junkers
ChemFiles 2008, 8.7, 3.
Keywords: Absorption, Addition reactions, Alkylations, Asymmetric synthesis, Building blocks, Degradations, Drug discovery, Metabolism, Molecular probes, Peptidomimetics, Pharmaceutical

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?