• USA Home
  • 672475 - 1-[4-(1H,1H,2H,2H-Perfluorodecyl)phenyl)-1,1-diphenylmethanol

EMAIL THIS PAGE TO A FRIEND
672475 Sigma-Aldrich

1-[4-(1H,1H,2H,2H-Perfluorodecyl)phenyl)-1,1-diphenylmethanol

98% (HPLC)

Synonym: 1-[4-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluordecyl)-phenyl]-1,1-diphenylmethanol, F17 Trityl OH

  • CAS Number 649561-66-0

  • Empirical Formula (Hill Notation) C29H19F17O

  • Molecular Weight 706.43

  •  MDL number MFCD09750443

  •  PubChem Substance ID 24885118

  •  NACRES NA.22

Purchase

Properties

Related Categories Chemical Synthesis, Fluorous Synthesis, Rf-Tagged Protecting Groups, Specialty Synthesis
Quality Level   100
assay   98% (HPLC)
mp   76-80 °C
SMILES string   OC(c1ccccc1)(c2ccccc2)c3ccc(CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)cc3
InChI   1S/C29H19F17O/c30-21(31,23(32,33)24(34,35)25(36,37)26(38,39)27(40,41)28(42,43)29(44,45)46)16-15-17-11-13-20(14-12-17)22(47,18-7-3-1-4-8-18)19-9-5-2-6-10-19/h1-14,47H,15-16H2
InChI key   ONXVYKQEANDRAX-UHFFFAOYSA-N

Description

Packaging

1 g in poly bottle

Legal Information

Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?