EMAIL THIS PAGE TO A FRIEND
672920 Sigma-Aldrich

TCA Deblock

3 % (w/v) in methylene chloride

Synonym: Deblock trichloroacetic acid solution

  • CAS Number 76-03-9

  • Empirical Formula (Hill Notation) C2HCl3O2

  • Molecular Weight 163.39

  •  MDL number MFCD00004177

  •  PubChem Substance ID 329760535

Purchase

Properties

Related Categories Chemical Synthesis, Deblock, Phosphoramidites and Reagents for Oligonucleotide Synthesis (Proligo Reagents), TCA Deblock with Dichloromethane
concentration   3 % (w/v) in methylene chloride
bp   39.8-40 °C (lit.)
density   1.327 g/mL at 25 °C (lit.)
SMILES string   OC(=O)C(Cl)(Cl)Cl
InChI   1S/C2HCl3O2/c3-2(4,5)1(6)7/h(H,6,7)
InChI key   YNJBWRMUSHSURL-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN2922 - class 8 - PG 3 - EHS - acidic - Corrosive liquids, toxic, n.o.s., HI: all
WGK Germany 
2
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?