• USA Home
  • 676039 - 5-Bromobenzo[1,3]dioxole-4-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
676039 Sigma-Aldrich

5-Bromobenzo[1,3]dioxole-4-carboxylic acid

97%

Purchase

Properties

Related Categories Building Blocks, C8, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
Quality Level   100
assay   97%
mp   169-173 °C
SMILES string   OC(=O)c1c(Br)ccc2OCOc12
InChI   1S/C8H5BrO4/c9-4-1-2-5-7(13-3-12-5)6(4)8(10)11/h1-2H,3H2,(H,10,11)
InChI key   HDNXEXSQANVCKC-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?