• USA Home
  • 676616 - trans-2-[3,5-Bis(trifluoromethyl)phenyl]vinylboronic acid pinacol ester

EMAIL THIS PAGE TO A FRIEND
676616 Sigma-Aldrich

trans-2-[3,5-Bis(trifluoromethyl)phenyl]vinylboronic acid pinacol ester

97%

Synonym: E-2-[3,5-Bis(trifluoromethyl)phenyl]vinylboronic acid pinacol ester

  • Empirical Formula (Hill Notation) C16H17BF6O2

  • Molecular Weight 366.11

  •  MDL number MFCD09038439

  •  PubChem Substance ID 24884858

Purchase

Properties

Related Categories Alkenyl Boronate Esters, Boronate Esters, Boronic Acids and Derivatives, Chemical Synthesis, Organometallic Reagents More...
assay   97%
mp   36-40 °C
SMILES string   [H]\C(B1OC(C)(C)C(C)(C)O1)=C(\[H])c2cc(cc(c2)C(F)(F)F)C(F)(F)F
InChI   1S/C16H17BF6O2/c1-13(2)14(3,4)25-17(24-13)6-5-10-7-11(15(18,19)20)9-12(8-10)16(21,22)23/h5-9H,1-4H3/b6-5+
InChI key   FNYGUDITBZMNGD-AATRIKPKSA-N

Description

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Boronic Acid Esters - Chemfiles Volume 4 Article 2

The synthesis of biaryl compounds via the Suzuki–Miyaura coupling reaction has become more commonplace now that many arylboronic acids are readily available. Several years ago, Miyaura et al. demonst...
Chemfiles Volume 4 Article 2
Keywords: Coupling reactions, Suzuki-Miyaura coupling

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?