• USA Home
  • 676683 - (–)-9-(1R,2R-Pseudoephedrinyl)-(10S)-(trimethylsilyl)-9-borabicyclo[3.3.2]decane

EMAIL THIS PAGE TO A FRIEND
676683 Aldrich

(–)-9-(1R,2R-Pseudoephedrinyl)-(10S)-(trimethylsilyl)-9-borabicyclo[3.3.2]decane

Purchase

Properties

Related Categories Allylation, Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents
InChI Key   ZOZYQBFBLCMBTA-MBMVPVEPSA-N
mp   135-148 °C

Description

Application

Air-stable, crystalline pseudoephedrine borinic acid complex which when treated with an appropriate Grignard reagent provides access to homoallylic or homopropargylic alcohols and α-allenyl carbinols.

Soderquist Reagents: For allylboration, allenylboration, propargylboration

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Soderquist Borabicyclodecanes

In the past 20 years, the asymmetric allylboration of aldehydes has become an essential reaction for the synthesis of homoallylic or homopropargylic alcohols. Soderquist et al. developed a robust, ve...
William Sommer
Aldrich ChemFiles 2008, 8.6, 8.
Keywords: Alkynylborations, Allylborations, Asymmetric synthesis, Chemfiles, Crotylborations, Grignard Reaction

Related Content

Soderquist Reagents

The Soderquist group at the University of Puerto Rico recently reported a number of versatile synthetic methods employing pseudoephedrine (PE)-derived borabicyclodecane auxiliaries 676675 and 676683 ...
Keywords: Aldrichimica Acta, Alkynylborations, Allylborations, Asymmetric synthesis, Crotylborations, Filtration, Grignard Reaction, Methods, Tools

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?