• USA Home
  • 67737 - Methyllithium solution

EMAIL THIS PAGE TO A FRIEND
67737 Sigma-Aldrich

Methyllithium solution

~1 M in cumene/THF

Synonym: Lithium methanide, MeLi

  • CAS Number 917-54-4

  • Empirical Formula (Hill Notation) CH3Li

  • Molecular Weight 21.98

  •  Beilstein/REAXYS Number 3587162

  •  MDL number MFCD00008253

  •  PubChem Substance ID 24885367

Purchase

Properties

concentration   ~1 M in cumene/THF
density   0.85 g/mL at 20 °C
storage temp.   2-8°C
SMILES string   [Li]C
InChI   1S/CH3.Li/h1H3;
InChI key   DVSDBMFJEQPWNO-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Signal word 
Danger
Supplemental Hazard Statements 
May form explosive peroxides.
RIDADR 
UN 3399A 4.3(3) / PGI
WGK Germany 
1
Flash Point(F) 
12.2 °F
Flash Point(C) 
-11 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Molarity Calculator & Normality Calculator for Acids & Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution. To ...

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?