• USA Home
  • 677612 - 4-Isobutylpyridine hydrochloride

EMAIL THIS PAGE TO A FRIEND
677612 Sigma-Aldrich

4-Isobutylpyridine hydrochloride

95%

Purchase

Properties

Related Categories Building Blocks, C9, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
assay   95%
mp   171-177 °C
SMILES string   Cl[H].CC(C)Cc1ccncc1
InChI   1S/C9H13N.ClH/c1-8(2)7-9-3-5-10-6-4-9;/h3-6,8H,7H2,1-2H3;1H
InChI key   KMABXTSPDGNDIA-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?