EMAIL THIS PAGE TO A FRIEND
678384 Sigma-Aldrich

6-Nitroquinoxaline

97%

  • CAS Number 6639-87-8

  • Empirical Formula (Hill Notation) C8H5N3O2

  • Molecular Weight 175.14

  •  MDL number MFCD00462822

  •  PubChem Substance ID 24885426

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Quinoxalines
InChI Key   YLKFDRWBZAALPN-UHFFFAOYSA-N
assay   97%
mp   175-179 °C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?