EMAIL THIS PAGE TO A FRIEND
67867 SAFC

Fmoc-Asn(Trt)-OPfp

≥98.0% (HPLC)

Synonym: N-α-Fmoc-N-β-trityl-L-asparagine pentafluorophenyl ester

  • CAS Number 132388-64-8

  • Empirical Formula (Hill Notation) C44H31F5N2O5

  • Molecular Weight 762.72

  •  MDL number MFCD00153373

Purchase

Properties

assay   ≥98.0% (HPLC)
optical activity   [α]/D -7.0±1.0°, c = 1 in dioxane
storage temp.   2-8°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?