• USA Home
  • 679194 - Pentyltrichlorosilane

EMAIL THIS PAGE TO A FRIEND
679194 Sigma-Aldrich

Pentyltrichlorosilane

deposition grade, ≥98%

Synonym: Amyltrichlorosilane

Purchase

Properties

Related Categories Chemical Synthesis, Materials Science, Micro/NanoElectronics, Protecting and Derivatizing Reagents, Protection and Derivatization,
grade   deposition grade
InChI Key   KWDQAHIRKOXFAV-UHFFFAOYSA-N
assay   ≥98%
form   liquid
refractive index   n20/D 1.4405 (lit.)
bp   173 °C(lit.)
density   1.142 g/mL at 25 °C (lit.)

Description

Packaging

50 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 2986 3(8) / PGII
WGK Germany 
1
RTECS 
VV4725000
Flash Point(F) 
87.8 °F
Flash Point(C) 
31 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Bio-Inspired “Green” Synthesis of Nanomaterials and their Applications

Dr. Siddharth V. Patwardhan Department of Chemical and Process Engineering University of Strathclyde, 75 Montrose Street Glasgow G1 1XJ, UK Email: Siddharth.Patwardhan@strath.ac.uk
Keywords: Adsorption, Agriculture, Apoptosis, Cancer, Catalysis, Condensations, Cosmetics, Deposition, Environmental, Enzyme activity, Green chemistry, Hydration reaction, Immobilization, Microwave synthesis, Nanomaterials, Pharmaceutical, Precipitation, Reductions, Separation, Sol-gel, Solvents

Deposition Grade Silanes for Sol-Gel Processes

Sol-gel processing is beneficial in the formation of ceramic and glass films for a number of reasons.1-4 It is a simple process that does not require exotic materials, catalysts or expensive depositi...
Keywords: Chromatography, Condensations, Deposition, Electronics, Gas chromatography, Gas chromatography mass spectrometry, Mass spectrometry, Nuclear magnetic resonance spectroscopy, Sol-gel

Synthesis of Melting Gels Using Mono-Substituted and Di-Substituted Alkoxysiloxanes

Hybrid organic-inorganic sol-gel materials containing silica were first called “ORMOSILs” in 1984.1 Since then, the number of hybrid organicinorganic combinations has increased rapidly.2 Hybrid mater...
Lisa C. Klein and Andrei Jitianu
Material Matters, Volume 7, Number 2, 8–14
Keywords: Absorption, Adhesion, Calorimetry, Combustion, Condensations, Deposition, Exothermic, Filtration, Materials Science, Melting, Oxidations, Polymerization reactions, Purification, Semiconductor, Sol-gel, Solar cells, Solvents, Spin coating, Substitutions, Thermal analysis

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?