EMAIL THIS PAGE TO A FRIEND
681776 SAFC

Phosgene solution

20% in toluene

  • CAS Number 75-44-5

  • Linear Formula COCl2

  • Molecular Weight 98.92

  •  Beilstein/REAXYS Number 1098367

  •  MDL number MFCD00036119

  •  PubChem Substance ID 24885654

Purchase

Properties

concentration   20% in toluene
density   0.935 g/mL at 25 °C
SMILES string   ClC(Cl)=O
InChI   1S/CCl2O/c2-1(3)4
InChI key   YGYAWVDWMABLBF-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Signal word 
Danger
Target organs 
Central nervous system
RIDADR 
UN3489 - class 6.1 - PG 1 - acidic - RQ - Toxic by inhalation liquid, flammable, corrosive, n.o.s., HI: all (not BR)
WGK Germany 
WGK 2
Flash Point(F) 
39.2 °F - closed cup
Flash Point(C) 
4 °C - closed cup

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?