• USA Home
  • 682853 - 3-Benzyloxybenzoyl chloride

EMAIL THIS PAGE TO A FRIEND
682853 Aldrich

3-Benzyloxybenzoyl chloride

95%

Synonym: 3-Benzyloxybenzoic acid chloride

  • CAS Number 61535-46-4

  • Empirical Formula (Hill Notation) C14H11ClO2

  • Molecular Weight 246.69

  •  MDL number MFCD02258040

Purchase

Properties

InChI Key   HPVUGOBQRQUWMK-UHFFFAOYSA-N
assay   95%
mp   40-43 °C

Description

Application

Reactant for:
• Preparation of dihydroquinolinones and tetrahydroquinazolinones as potent kinesin spindle protein inhibitors and potential antitumor agents
• Preparation of carbamate derivatives as fatty acid amide hydrolase inhibitors

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN3261 - class 8 - PG 2 - EHS - Corrosive solid, acidic, organic, n.o.s., HI: all
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?