• USA Home
  • 684376 - 1,3-Bis[(phenylseleno)methyl]benzene

EMAIL THIS PAGE TO A FRIEND
684376 Sigma-Aldrich

1,3-Bis[(phenylseleno)methyl]benzene

97%

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Other Ligands More...
Quality Level   100
assay   97%
reaction suitability   reagent type: ligand
mp   38-42 °C
SMILES string   C([Se]c1ccccc1)c2cccc(C[Se]c3ccccc3)c2
InChI   1S/C20H18Se2/c1-3-10-19(11-4-1)21-15-17-8-7-9-18(14-17)16-22-20-12-5-2-6-13-20/h1-14H,15-16H2
InChI key   KNRBPZAUCBNUPW-UHFFFAOYSA-N

Description

Application

Reactant for synthesis of:
• Allylic and benzylic carbanions
• SeCSe-Pd(II) pincer complex
• Palladium catalysts for the Heck reaction

Precursor ligand for preparation of Se-ligated Pd-pincer catalyst.

Packaging

1 g in glass bottle

250 mg in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3283 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Catalysts
Protocols & Articles

Articles

Metal and Phosphine Mediated Transformations - Aldrich ChemFiles 2009, 9.2, 7.

Functionalized allyl boronates are useful building blocks in natural product synthesis. Olsson et al. reported the use of a palladium pincer complex in combination with diboronic acid for the boronat...
Josephine Nakhla
Aldrich ChemFiles 2009, 9.2, 7.
Keywords: Building blocks, Chemfiles, Esterifications, Natural product synthesis

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?