• USA Home
  • 685895 - N,N-Diethyl-2-(trimethylsilyl)indole-1-carboxamide

EMAIL THIS PAGE TO A FRIEND
685895 Sigma-Aldrich

N,N-Diethyl-2-(trimethylsilyl)indole-1-carboxamide

90%

Purchase

Properties

Related Categories Building Blocks, C13 to C27, Chemical Synthesis, Heterocyclic Building Blocks, Indoles More...
assay   90%
refractive index   n20/D 1.555
density   1.030 g/mL at 25 °C
SMILES string   CCN(CC)C(=O)n1c(cc2ccccc12)[Si](C)(C)C
InChI   1S/C16H24N2OSi/c1-6-17(7-2)16(19)18-14-11-9-8-10-13(14)12-15(18)20(3,4)5/h8-12H,6-7H2,1-5H3
InChI key   FVRCYZLAJZNZRQ-UHFFFAOYSA-N

Description

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Indoles, Purines, and Their Isosteres

Substituted indoles and purines have frequently been referred to as “privileged structures” since they are capable of binding to multiple receptors with high affinity, and thus have applications acro...
Keywords: Building blocks, Medicinal chemistry

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?