EMAIL THIS PAGE TO A FRIEND
685925 Sigma-Aldrich

(S,S)-f-BINAPHANE

Synonym: 1,1′-Bis[(11bs)-3,5-dihydro-4H-dinaphtho[2,1-c:1′,2′-e]phosphenpin-4yl]ferrocene, 1,1′-Bis[(S)-4,5-dihydro-3H-binaphtho[2,1-c:1′,2′-e]phosphepino]ferrocene

  • CAS Number 544461-38-3

  • Empirical Formula (Hill Notation) C54H40P2Fe

  • Molecular Weight 806.69

  •  MDL number MFCD09842757

Purchase

Properties

SMILES string   [Fe].[CH]1[CH][CH][C]([CH]1)P2Cc3ccc4ccccc4c3-c5c(C2)ccc6ccccc56.[CH]7[CH][CH][C]([CH]7)P8Cc9ccc%10ccccc%10c9-c%11c(C8)ccc%12ccccc%11%12

Description

Legal Information

Sold in collaboration with Chiral Quest for research purposes only. US Patent No.: 6,828,271.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

ChiralQuest Phosphine Ligands and Complexes

Professor Xumu Zhang at Penn State has made remarkable advances by creating a toolbox of chiral phosphines which can be used on a variety of substrates, some of which have been historically resistant...
William Sommer and Daniel Weibel
Aldrich ChemFiles 2008, 8.2, 47.
Keywords: Asymmetric catalysis, Asymmetric synthesis, Catalysis, Chemfiles, Cyclizations, Hydrogenations, Ligands, Reductions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?