• USA Home
  • 686913 - 4-(4-Bromophenyl)thiophene-2-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
686913 Sigma-Aldrich

4-(4-Bromophenyl)thiophene-2-carboxylic acid

97%

Purchase

Properties

Related Categories Building Blocks, C10 to C20+, Chemical Synthesis, Heterocyclic Building Blocks, Thiophenes More...
InChI Key   PZHUPHYFDBYWCQ-UHFFFAOYSA-N
assay   97%
mp   238-242 °C

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?