• USA Home
  • 688819 - trans-2-(Aminomethyl)cyclohexanol hydrochloride

EMAIL THIS PAGE TO A FRIEND
688819 Sigma-Aldrich

trans-2-(Aminomethyl)cyclohexanol hydrochloride

97%

  • CAS Number 24948-05-8

  • Empirical Formula (Hill Notation) C7H15NO · HCl

  • Molecular Weight 165.66

  •  Beilstein/REAXYS Number 3674924

  •  MDL number MFCD00145420

  •  PubChem Substance ID 329761196

Purchase

Properties

Related Categories Amino Alcohols, Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds More...
assay   ≥96.5% (GC)
  96.5-103.5% (AT)
  97%
storage temp.   2-8°C
SMILES string   Cl.NC[C@@H]1CCCC[C@H]1O
InChI   1S/C7H15NO.ClH/c8-5-6-3-1-2-4-7(6)9;/h6-7,9H,1-5,8H2;1H/t6-,7+;/m0./s1
InChI key   UVYXWKUCIQHOCP-UOERWJHTSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?