• USA Home
  • 689653 - O-(2-Carboxyethyl)-O′-[2-(Fmoc-amino)-ethyl]heptacosaethylene glycol

EMAIL THIS PAGE TO A FRIEND
689653 Sigma-Aldrich

O-(2-Carboxyethyl)-O′-[2-(Fmoc-amino)-ethyl]heptacosaethylene glycol

≥90% (oligomer purity)

Synonym: Fmoc-amino PEG propionic acid (n = 27)

  • Empirical Formula (Hill Notation) C74H129NO32

  • Molecular Weight 1544.80

  •  PubChem Substance ID 329761280

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Heterobifunctional PEG, PEGylation
Quality Level   100
assay   ≥90% (oligomer purity)
mol wt   average Mn 1,600
reaction suitability   reagent type: cross-linking reagent
Ω-end   Fmoc
α-end   carboxylic acid
storage temp.   −20°C
SMILES string   OC(=O)CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCCCNC(=O)OCC1c2ccccc2-c3ccccc13
InChI   1S/C76H133NO32/c78-75(79)11-14-82-16-18-84-20-22-86-24-26-88-28-30-90-32-34-92-36-38-94-40-42-96-44-46-98-48-50-100-52-54-102-56-58-104-60-62-106-64-66-108-68-67-107-65-63-105-61-59-103-57-55-101-53-51-99-49-47-97-45-43-95-41-39-93-37-35-91-33-31-89-29-27-87-25-23-85-21-19-83-17-15-81-13-6-5-12-77-76(80)109-69-74-72-9-3-1-7-70(72)71-8-2-4-10-73(71)74/h1-4,7-10,74H,5-6,11-69H2,(H,77,80)(H,78,79)
InChI key   WTZSFCYBPNRWRK-UHFFFAOYSA-N

Description

Packaging

100 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Cell Culture Scaffolds via Bio-orthogonal Click Reactions

1,2Malar A. Azagarsamy, 1Navakanth R. Gandavarapu, 1,2Kristi S. Anseth* 1Department of Chemical and Biological Engineering and 2the Howard Hughes Medical Institute, University of Colorado at Boulder,...
Keywords: Addition reactions, Adhesion, Biomaterials, Catalysis, Cell culture, Cell signaling, Chemical reactions, Click chemistry, Cycloadditions, Degradations, Growth factors, Huisgen Cycloaddition, Ligands, Michael Addition, Peptide synthesis, Polymerization reactions, Step-growth polymerization

Degradable Poly(ethylene glycol) Hydrogels for 2D and 3D Cell Culture

Progress in biotechnology fields such as tissue engineering and drug delivery is accompanied by an increasing demand for diverse functional biomaterials. For decades, research in polymeric biomateria...
Keywords: Absorption, Angiogenesis, Biomaterials, Catalysis, Cell culture, Degradations, Eliminations, Growth factors, Infrared spectroscopy, Ligands, Michael Addition, Polymerization reactions, Ring-opening polymerization

Polyethylene Glycol (PEG) Selection Guide

Poly(ethylene glycol) (PEG) is a synthetic, hydrophilic, biocompatible polymer with widespread use in biomedical and other applications. PEGs are synthesized using a ring-opening polymerization of et...
Keywords: Adhesion, Bacterial conjugations, Click chemistry, Molecular biology, PEGylations, Polymerization reactions, Ring-opening polymerization

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?