EMAIL THIS PAGE TO A FRIEND
690147 Sigma-Aldrich

1-Ethylimidazole

produced by BASF, ≥95.0% (HPLC)

Purchase

Properties

Related Categories Building Blocks, C3 to C8, Chemical Synthesis, Heterocyclic Building Blocks, Imidazoles,
grade   produced by BASF
InChI Key   IWDFHWZHHOSSGR-UHFFFAOYSA-N
assay   ≥95.0% (HPLC)
impurities   ≤0.5% water
color   APHA: ≤100

Description

Caution

product may discolour on storage

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3267 8 / PGIII
WGK Germany 
2
Flash Point(F) 
195.8 °F
Flash Point(C) 
91 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?