• USA Home
  • 691607 - 4-Bromobutylboronic acid

EMAIL THIS PAGE TO A FRIEND
691607 Sigma-Aldrich

4-Bromobutylboronic acid

≥95%

  • CAS Number 61632-72-2

  • Empirical Formula (Hill Notation) C4H10BBrO2

  • Molecular Weight 180.84

  •  MDL number MFCD10000895

  •  PubChem Substance ID 329761423

  •  NACRES NA.22

Purchase

Properties

Related Categories Alkyl and Aryl Boronic Acids, Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Organometallic Reagents More...
Quality Level   100
assay   ≥95%
mp   73-83 °C
storage temp.   2-8°C
SMILES string   OB(O)CCCCBr
InChI   1S/C4H10BBrO2/c6-4-2-1-3-5(7)8/h7-8H,1-4H2
InChI key   VYJXKSGBEKVCJK-UHFFFAOYSA-N

Description

Application

Reactant for:
• Stereoselective preparation of chiral tertiary alkylamines via amination with tetrachlorosilane and alkyl azides

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?