• USA Home
  • 691798 - Vanadium(III) bromide

EMAIL THIS PAGE TO A FRIEND
691798 Sigma-Aldrich

Vanadium(III) bromide

99%

Purchase

Properties

Related Categories Alternative Energy, Catalysts, Materials Science, Materials for Hydrogen Storage
assay   99%
density   4 g/mL at 25 °C (lit.)
SMILES string   Br[V](Br)Br
InChI   1S/3BrH.V/h3*1H;/q;;;+3/p-3
InChI key   ZOYIPGHJSALYPY-UHFFFAOYSA-K

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3260 8 / PGII
WGK Germany 
3
RTECS 
YW2750000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?