• USA Home
  • 692506 - (R)-RuCl[(p-cymene)(T-BINAP)]Cl

EMAIL THIS PAGE TO A FRIEND
692506 Sigma-Aldrich

(R)-RuCl[(p-cymene)(T-BINAP)]Cl

Synonym: Chloro[(R)-(+)-2,2′-bis(di-p-tolylphosphino)-1,1′-binaphthyl](p-cymene)ruthenium(II) chloride

  • CAS Number 131614-43-2

  • Linear Formula [C58H54ClP2Ru]+Cl-

  • Molecular Weight 984.97

  •  PubChem Substance ID 329761503

Purchase

Properties

Related Categories Asymmetric Synthesis, BINAPs, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Privileged Ligands and Complexes More...
InChI Key   BHIUOOHVIWKUTN-UHFFFAOYSA-L
storage temp.   2-8°C

Description

Legal Information

Sold in collaboration with Takasago for research purposes only.

Application

Takasago Ligands and Complexes for Asymmetric Reactions

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Takasago Ligands and Complexes for Asymmetric Reactions

The quest to find chiral catalysts capable of achieving high enantiomeric excess for a variety of asymmetric transformations has been an ongoing endeavor for the past 40 years. One of the most releva...
Keywords: Addition reactions, Arylations, Asymmetric synthesis, Catalysis, Hydrogenations, Ligands, transformation

Related Content

Takasago Ligands - Advantages and Applications

Introduction The quest to find chiral catalysts capable of achieving high enantiomeric excess for a variety of asymmetric transformations has been an ongoing endeavor for the past 40 years. One of th...
Keywords: Addition reactions, Applications, Arylations, Asymmetric synthesis, Building blocks, Catalysis, Epoxidations, Hydrogenations, Hydrosilylations, Ligands, Sharpless Epoxidation, transformation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?