• USA Home
  • 693170 - (S)-[(RuCl(DM-SEGPHOS®))2(μ-Cl)3][NH2Me2]

EMAIL THIS PAGE TO A FRIEND
693170 Sigma-Aldrich

(S)-[(RuCl(DM-SEGPHOS®))2(μ-Cl)3][NH2Me2]

Synonym: Dimethylammonium dichlorotri(μ-chloro)bis[(S)-(−)-5,5′-bis[di(3,5-xylyl)phosphino]-4,4′-bi-1,3-benzodioxole]diruthenate(II)

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Privileged Ligands and Complexes, SEGPHOS More...
InChI Key   YOYDPZZYORNKOE-UHFFFAOYSA-M
storage temp.   2-8°C

Description

Application

Takasago Ligands and Complexes for Asymmetric Reactions

Packaging

50, 100 mg in glass bottle

Legal Information

Sold in collaboration with Takasago for research purposes only. JP Registration No. 3148136, 3549390

SEGPHOS is a registered trademark of Takasago Intl. Corp.

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN1325 - class 4.1 - PG 3 - Flammable solids, organic, n.o.s., HI: all
WGK Germany 
3

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles

Articles

Takasago Ligands and Complexes for Asymmetric Reactions

The quest to find chiral catalysts capable of achieving high enantiomeric excess for a variety of asymmetric transformations has been an ongoing endeavor for the past 40 years. One of the most releva...
Keywords: Addition reactions, Arylations, Asymmetric synthesis, Catalysis, Hydrogenations, Ligands, transformation

Related Content

Takasago Ligands - Advantages and Applications

Introduction The quest to find chiral catalysts capable of achieving high enantiomeric excess for a variety of asymmetric transformations has been an ongoing endeavor for the past 40 years. One of th...
Keywords: Addition reactions, Applications, Arylations, Asymmetric synthesis, Building blocks, Catalysis, Epoxidations, Hydrogenations, Hydrosilylations, Ligands, Sharpless Epoxidation, transformation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?